Gene Transcripts

Gene Symbol
1110064A23Rik
Entrez Gene ID
68893
Gene Name
RIKEN cDNA 1110064A23 gene
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Mm.69566
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK004365.1 - 1 74
BC040515.1 - 1 1137
BF180451.1 - 1 167


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BY704595.1 - 1 74 Chr. 2 - 31948547 - 31949083 [+]
GenBankmRNA CF539586.1 - 1 72 Chr. 2 - 31948549 - 31949081 [+]

View Full Product Details