Gene Transcripts
- Gene Symbol
- LINC01551
- Entrez Gene ID
- 387978
- Gene Name
- long intergenic non-protein coding RNA 1551
- Gene Aliases
- C14orf23, c14_5148
- Chromosome Location
- Chr. 14: 28772704 - 28794794 on Build GRCh38
- UniGene ID
- Hs.633254
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026731.1 |
|
3 |
1553 |
|
View Full Product Details