Gene Transcripts

Gene Symbol
ARHGEF26-AS1
Entrez Gene ID
100507524
Gene Name
ARHGEF26 antisense RNA 1
Gene Aliases
Chromosome Location
Chr. 3: 154024401 - 154121210 on Build GRCh38
UniGene ID
Hs.370221
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_037901.1 3 336
GenBank mRNA BC038431.1 - 3 338


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GENCODE ENST00000491862.5 - 3 336 Chr. 3 - 154024401 - 154121210 [-]
GENCODE ENST00000480639.5 - 3 398 Chr. 3 - 154026479 - 154121263 [-]

View Full Product Details