Gene Transcripts
- Gene Symbol
- Ammecr1l
- Entrez Gene ID
- 225339
- Gene Name
- AMME chromosomal region gene 1-like
- Gene Aliases
- 5430429D03Rik, AU022236, AU040755, AW111353, E230022H04Rik
- Chromosome Location
- Chr. 18: 31759475 - 31784083 on Build GRCm38
- UniGene ID
- Mm.290804
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK041287.1 |
- |
|
363 |
|
View Full Product Details