Gene Transcripts

Gene Symbol
C22orf24
Entrez Gene ID
25775
Gene Name
chromosome 22 open reading frame 24
Gene Aliases
HSN44A4A
Chromosome Location
Chr. 22: 31933521 - 31945362 on Build GRCh38
UniGene ID
Hs.627602
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001302819.1 NP_001289748.1 1 347
NM_001302820.1 NP_001289749.1 1 347
GenBank mRNA AK294232.1 - 1 317
DA485290.1 - 1 348

View Full Product Details