Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_176850.2 | NP_789820.2 | 3 | 1631 | |
GenBank mRNA | BC003956.1 | - | 2 | 931 | |
BC037661.1 | - | 3 | 1276 | ||
BC046973.1 | - | 3 | 1346 | ||
CN724595.1 | - | 1 | 35 | ||
DV660897.1 | - | 2 | 664 | ||
GU189417.1 | - | 3 | 1184 |