Gene Transcripts

Gene Symbol
Armc2
Entrez Gene ID
213402
Gene Name
armadillo repeat containing 2
Gene Aliases
2610018I05Rik
Chromosome Location
Chr. 10: 41907755 - 42018382 on Build GRCm38
UniGene ID
Mm.211320
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001034858.3 NP_001030030.2 7, 8 1046
GenBank mRNA AK131686.1 - 3, 4 241
AK141039.1 - 1, 2 1378
BC172168.1 - 1, 2 756

View Full Product Details