Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_153800.4 | NP_722495.3 | 9, 10 | 1146 | |
GenBank mRNA | AK046097.1 | - | 9, 10 | 1148 | |
AK052541.1 | - | 6, 7 | 792 | ||
AK078218.1 | - | 8, 9 | 1035 | ||
AY541447.1 | - | 5, 6 | 979 | ||
BC038272.1 | - | 4, 5 | 644 | 4237331 |