Gene Transcripts

Gene Symbol
Cachd1
Entrez Gene ID
320508
Gene Name
cache domain containing 1
Gene Aliases
1190007F10Rik, AI852726, B430218L07Rik, Vwcd1, mKIAA1573
Chromosome Location
Chr. 4: 100776679 - 101003749 on Build GRCm38
UniGene ID
Mm.151962
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198037.1 NP_932154.1 27 4109
GenBank mRNA BC057356.1 - 2395
BC058676.1 - 27 4109 6834428
BC080313.1 - 2393

View Full Product Details