Gene Transcripts

Gene Symbol
Btbd6
Entrez Gene ID
399566
Gene Name
BTB (POZ) domain containing 6
Gene Aliases
Chromosome Location
Chr. 12: 112974998 - 112979493 on Build GRCm38
UniGene ID
Mm.440140
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145900.1 NP_001139372.1 5 755
NM_201646.2 NP_964008.2 4 837
GenBank mRNA AK146991.1 - 4 669
BC031195.1 - 4 837 5059510

View Full Product Details