Gene Transcripts

Gene Symbol
Baz2b
Entrez Gene ID
407823
Gene Name
bromodomain adjacent to zinc finger domain, 2B
Gene Aliases
5830435C13Rik, BC053917, D2Ertd794e
Chromosome Location
Chr. 2: 59899363 - 60209846 on Build GRCm38
UniGene ID
Mm.228071
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001182.3 NP_001001182.2 20 3197
GenBank mRNA AK029231.1 - 14 2245
AK173180.1 - 20 3229
BC150814.1 - 22 3206 9087725

View Full Product Details