Gene Transcripts
- Gene Symbol
- Cast
- Entrez Gene ID
- 25403
- Gene Name
- calpastatin
- Gene Aliases
- Chromosome Location
- Chr. 2: 1451933 - 1561666 on Build Rnor_6.0
- UniGene ID
- Rn.17481
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001033715.1 |
NP_001028887.1
|
18, 19 |
1501 |
|
NM_001033716.1 |
NP_001028888.1
|
17, 18 |
1432 |
|
NM_053295.2 |
NP_445747.2
|
19, 20 |
1615 |
|
XM_006231698.2 |
XP_006231760.1
|
19, 20 |
2023 |
|
XM_006231699.2 |
XP_006231761.1
|
18, 19 |
1748 |
|
GenBank mRNA |
AF346597.1 |
- |
15, 16 |
1113 |
|
AF346598.1 |
- |
17, 18 |
1296 |
|
BC091239.1 |
- |
18, 19 |
1501 |
7324026 |
DQ186624.1 |
- |
19, 20 |
1809 |
|
DQ186625.1 |
- |
17, 18 |
1626 |
|
DQ186626.1 |
- |
18, 19 |
1740 |
|
DQ186627.1 |
- |
18, 19 |
1695 |
|
DQ186628.1 |
- |
17, 18 |
1656 |
|
DQ186629.1 |
- |
16, 17 |
1587 |
|
DQ287975.1 |
- |
12, 13 |
1487 |
|
Y13587.1 |
- |
16, 17 |
1194 |
|
Y13588.1 |
- |
17, 18 |
1308 |
|
Y13589.1 |
- |
15, 16 |
1125 |
|
View Full Product Details