Gene Transcripts

Gene Symbol
Chp1
Entrez Gene ID
64152
Gene Name
calcineurin-like EF-hand protein 1
Gene Aliases
Chp, Wrch2
Chromosome Location
Chr. 3: 111354506 - 111389998 on Build Rnor_6.0
UniGene ID
Rn.11041
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024139.2 NP_077053.1 4 467
GenBank mRNA AB070350.1 - 4 452
BC062029.1 - 4 467 5623922
U39875.1 - 4 713

View Full Product Details