Gene Transcripts

Gene Symbol
Arhgap36
Entrez Gene ID
302819
Gene Name
Rho GTPase activating protein 36
Gene Aliases
RGD1565967
Chromosome Location
Chr. X: 136466638 - 136495646 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006257554.2 XP_006257616.2 5 880
XM_006257555.2 XP_006257617.1 5 903
XM_229133.8 XP_229133.5 5 880

View Full Product Details