Gene Transcripts

Gene Symbol
Astn1
Entrez Gene ID
304900
Gene Name
astrotactin 1
Gene Aliases
Chromosome Location
Chr. 13: 76055255 - 76370674 on Build Rnor_6.0
UniGene ID
Rn.28132
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001170603.1 NP_001164074.1 8 1809
XM_008769640.1 XP_008767862.1 5 1227
XM_008769641.1 XP_008767863.1 5 1202

View Full Product Details