Gene Transcripts
- Gene Symbol
- ARF5
- Entrez Gene ID
- 381
- Gene Name
- ADP ribosylation factor 5
- Gene Aliases
- Chromosome Location
- Chr. 7: 127588352 - 127591705 on Build GRCh38
- UniGene ID
- Hs.653202
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001662.3 |
NP_001653.1
|
2, 3 |
296 |
|
GenBank mRNA |
AF493884.1 |
- |
2, 3 |
149 |
|
AK297026.1 |
- |
2, 3 |
201 |
|
AK307297.1 |
- |
2, 3 |
270 |
|
AK308548.1 |
- |
2, 3 |
478 |
|
AK312107.1 |
- |
2, 3 |
201 |
|
BC003043.1 |
- |
2, 3 |
186 |
2822973 |
BC033104.1 |
- |
2, 3 |
243 |
4862014 |
BT007087.1 |
- |
2, 3 |
149 |
|
CN259625.1 |
- |
2, 3 |
313 |
|
M57567.1 |
- |
2, 3 |
203 |
|
View Full Product Details