Gene Transcripts

Gene Symbol
Arfgef1
Entrez Gene ID
312915
Gene Name
ADP-ribosylation factor guanine nucleotide-exchange factor 1(brefeldin A-inhibited)
Gene Aliases
BIG1, RGD1560793
Chromosome Location
Chr. 5: 8665614 - 8760458 on Build Rnor_6.0
UniGene ID
Rn.6834
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001277056.1 NP_001263985.1 16 2460
XM_008763497.1 XP_008761719.1 17 2869

View Full Product Details