Gene Transcripts

Gene Symbol
Cars2
Entrez Gene ID
361184
Gene Name
cysteinyl-tRNA synthetase 2, mitochondrial
Gene Aliases
RGD1311612
Chromosome Location
Chr. 16: 83288216 - 83325706 on Build Rnor_6.0
UniGene ID
Rn.54352
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006253460.2 XP_006253522.1 1 314
XM_006253461.2 XP_006253523.1 1 312
XM_006253462.2 XP_006253524.1 1 305

View Full Product Details