Gene Transcripts

Gene Symbol
Asap2
Entrez Gene ID
362719
Gene Name
ArfGAP with SH3 domain, ankyrin repeat and PH domain 2
Gene Aliases
Ddef2
Chromosome Location
Chr. 6: 43229927 - 43344332 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008764624.1 XP_008762846.1 17 6335
XM_008764625.1 XP_008762847.1 17 6335
XM_008764626.1 XP_008762848.1 16 6326

View Full Product Details