Gene Transcripts

Gene Symbol
ASH2L
Entrez Gene ID
9070
Gene Name
ASH2 like histone lysine methyltransferase complex subunit
Gene Aliases
ASH2, ASH2L1, ASH2L2, Bre2
Chromosome Location
Chr. 8: 38104965 - 38140080 on Build GRCh38
UniGene ID
Hs.521530
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105214.2 NP_001098684.1 10 1163
NM_001261832.1 NP_001248761.1 10 1163
NM_001282272.1 NP_001269201.1 9 916
NM_004674.4 NP_004665.2 10 1062
XM_005273682.1 XP_005273739.1 11 1608
XM_005273683.1 XP_005273740.1 11 1181
XM_006716412.1 XP_006716475.1 11 1608
XM_006716413.2 XP_006716476.1 10 1028
GenBank mRNA AB020982.1 - 10 1015
AF056717.1 - 10 1017
AF056718.1 - 10 1008
AK291750.1 - 10 1026
AK291938.1 - 10 1034
AK298483.1 - 9 859
AK311336.1 - 10 940
BC015936.2 - 10 1030 3921999

View Full Product Details