Gene Transcripts

Gene Symbol
Cbln3
Entrez Gene ID
501998
Gene Name
cerebellin 3 precursor
Gene Aliases
RGD1563731
Chromosome Location
Chr. 15: 34548113 - 34550940 on Build Rnor_6.0
UniGene ID
Rn.145788
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109330.1 NP_001102800.1 2 564
XM_006252007.2 XP_006252069.1 2 654

View Full Product Details