Gene Transcripts

Gene Symbol
Arid3c
Entrez Gene ID
502946
Gene Name
AT rich interactive domain 3C (BRIGHT-like)
Gene Aliases
RGD1560943
Chromosome Location
Chr. 5: 58107220 - 58114632 on Build Rnor_6.0
UniGene ID
Rn.225882
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001173981.1 NP_001167452.1 5 993
XM_008763641.1 XP_008761863.1 6 1207
XM_008763642.1 XP_008761864.1 6 1207

View Full Product Details