Gene Transcripts

Gene Symbol
Ccdc27
Entrez Gene ID
679584
Gene Name
coiled-coil domain containing 27
Gene Aliases
Chromosome Location
Chr. 5: 171312790 - 171327581 on Build Rnor_6.0
UniGene ID
Rn.231435
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191063.1 NP_001177992.1 7 1025
XM_008764384.1 XP_008762606.1 8 1189

View Full Product Details