Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004886.3 | NP_004877.1 | 11 | 2000 | |
XM_006722950.3 | XP_006723013.1 | 10 | 2126 | ||
XM_006722951.3 | XP_006723014.1 | 8 | 1811 | ||
GenBank mRNA | AB021638.1 | - | 11 | 1990 | |
AB209603.1 | - | 4 | 4199 | ||
BC008338.1 | - | 9 | 1847 | ||
BC086306.1 | - | 10 | 1859 | 6371716 |