Gene Transcripts

Gene Symbol
VPS9D1
Entrez Gene ID
9605
Gene Name
VPS9 domain containing 1
Gene Aliases
ATP-BL, C16orf7
Chromosome Location
Chr. 16: 89707133 - 89721046 on Build GRCh38
UniGene ID
Hs.164410
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004913.2 NP_004904.2 10 1372
XM_005256329.4 XP_005256386.1 10 1432
XM_005256330.4 XP_005256387.1 10 1249
XM_005256331.2 XP_005256388.1 7 906
XM_006721350.2 XP_006721413.1 10 1432
XM_011523476.2 XP_011521778.1 10 1429
XM_011523477.2 XP_011521779.1 9 3245
XM_011523478.2 XP_011521780.1 9 3242
XM_011523479.2 XP_011521781.1 10 1432
XM_011523480.2 XP_011521782.1 10 1286
GenBank mRNA AB018551.1 - 9 1412
BM920493.1 - 1 237

View Full Product Details