Gene Transcripts

Gene Symbol
C8G
Entrez Gene ID
733
Gene Name
complement component 8, gamma polypeptide
Gene Aliases
C8C
Chromosome Location
Chr. 9: 136944870 - 136946981 on Build GRCh38
UniGene ID
Hs.1285
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000606.2 NP_000597.2 4 517
GenBank mRNA BC113624.1 - 4 492
BC113626.1 - 4 492 8322678
BX282392.1 - 3 255
M17263.1 - 4 468
M17999.1 - 4 503
X06465.1 - 4 503

View Full Product Details