Gene Transcripts

Gene Symbol
ASPSCR1
Entrez Gene ID
79058
Gene Name
ASPSCR1, UBX domain containing tether for SLC2A4
Gene Aliases
ASPCR1, ASPL, ASPS, RCC17, TUG, UBXD9, UBXN9
Chromosome Location
Chr. 17: 81977550 - 82018929 on Build GRCh38
UniGene ID
Hs.298351
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024083.3 NP_076988.1 12, 13 1450
NR_045351.1 11, 12 1394
XM_011523601.2 XP_011521903.1 12, 13 1450
XM_011523602.2 XP_011521904.1 12, 13 1450
XM_017025036.1 XP_016880525.1 12, 13 1525
XM_017025037.1 XP_016880526.1 12, 13 1525
XM_017025038.1 XP_016880527.1 13, 14 1536
XM_017025039.1 XP_016880528.1 13, 14 1453
XM_017025040.1 XP_016880529.1 12, 13 1390
XM_017025041.1 XP_016880530.1 11, 12 1393
XM_017025042.1 XP_016880531.1 12, 13 1525
XM_017025043.1 XP_016880532.1 11, 12 1393
XM_017025044.1 XP_016880533.1 13, 14 1461
GenBank mRNA AF324219.2 - 12, 13 1375
AK057851.1 - 11, 12 1377
AK090682.1 - 5, 6 2815
AK126967.1 - 5, 6 5110
AK127159.1 - 5, 6 2022
AK226086.1 - 12, 13 1323
AK290624.1 - 12, 13 1366
BC006152.1 - 13, 14 1525 3357489
BC018722.1 - 12, 13 1355 4640076

View Full Product Details