Gene Transcripts

Gene Symbol
ARMC2
Entrez Gene ID
84071
Gene Name
armadillo repeat containing 2
Gene Aliases
bA787I22.1
Chromosome Location
Chr. 6: 108848410 - 108990738 on Build GRCh38
UniGene ID
Hs.645481
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005267154.3 XP_005267211.1 1 264

View Full Product Details