Gene Transcripts

Gene Symbol
Cpne2
Entrez Gene ID
234577
Gene Name
copine II
Gene Aliases
3322401K10Rik
Chromosome Location
Chr. 8: 94533028 - 94570529 on Build GRCm38
UniGene ID
Mm.291815
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153507.2 NP_705727.1 5 700
GenBank mRNA AK160238.1 - 5 690
BC022128.1 - 5 661 5097527
BC031801.1 - 5 700 3985959

View Full Product Details