Gene Transcripts
- Gene Symbol
- C8B
- Entrez Gene ID
- 732
- Gene Name
- complement component 8, beta polypeptide
- Gene Aliases
- C82
- Chromosome Location
- Chr. 1: 56929210 - 56974383 on Build GRCh38
- UniGene ID
- Hs.391835
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000066.3 |
NP_000057.2
|
6, 7 |
1055 |
|
NM_001278543.1 |
NP_001265472.1
|
7, 8 |
1277 |
|
NM_001278544.1 |
NP_001265473.1
|
7, 8 |
1184 |
|
XM_017002235.1 |
XP_016857724.1
|
7, 8 |
1123 |
|
GenBank mRNA |
AK292740.1 |
- |
6, 7 |
910 |
|
AK298425.1 |
- |
7, 8 |
1101 |
|
AK298444.1 |
- |
7, 8 |
1277 |
|
AK313382.1 |
- |
6, 7 |
927 |
|
AK316209.1 |
- |
7, 8 |
1274 |
|
BC020701.1 |
- |
6, 7 |
930 |
|
BC130575.1 |
- |
6, 7 |
888 |
40146606 |
M16973.1 |
- |
6, 7 |
890 |
|
X04393.1 |
- |
5, 6 |
725 |
|
View Full Product Details