Gene Transcripts

Gene Symbol
Muc15
Entrez Gene ID
690914
Gene Name
mucin 15, cell surface associated
Gene Aliases
Chromosome Location
Chr. 3: 101897782 - 101912979 on Build Rnor_6.0
UniGene ID
Rn.210834
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006234695.2 XP_006234757.1 5 1793
XM_008762107.1 XP_008760329.1 4 1570
XM_008762108.1 XP_008760330.1 4 1606

View Full Product Details