Gene Transcripts

Gene Symbol
CPM
Entrez Gene ID
1368
Gene Name
carboxypeptidase M
Gene Aliases
Chromosome Location
Chr. 12: 68842200 - 68963240 on Build GRCh38
UniGene ID
Hs.654387
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005502.2 NP_001005502.1 4, 5 473
NM_001874.4 NP_001865.1 4, 5 505
NM_198320.3 NP_938079.1 4, 5 491
GenBank mRNA AF368463.1 - 4, 5 463
AK130146.1 - 1, 2 81
AK313180.1 - 4, 5 463
BC022276.1 - 4, 5 463 4768771
CB959900.1 - 490
CB989009.1 - 4, 5 561
CD109360.1 - 4, 5 490
DA205368.1 - 4, 5 492
DA833423.1 - 1, 2 32
DB006895.1 - 4, 5 476
J04970.2 - 4, 5 505

View Full Product Details