Gene Transcripts

Gene Symbol
GAGE7
Entrez Gene ID
2579
Gene Name
G antigen 7
Gene Aliases
AL4, CT4.7, GAGE-7
Chromosome Location
Chr. : - on Build
UniGene ID
Hs.460641
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021123.2 NP_066946.1 5 445
GenBank mRNA AF058988.1 - 5 471

View Full Product Details