Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001128080.2 | NP_001121552.1 | 9 | 1206 | |
NM_001128081.1 | NP_001121553.1 | 7 | 1237 | ||
NM_001128082.1 | NP_001121554.1 | 6 | 1135 | ||
NM_020286.3 | NP_064682.1 | 8 | 1321 | ||
GenBank mRNA | AF175771.1 | - | 8 | 1170 | |
AF291425.1 | - | 8 | 1289 | ||
AJ279791.1 | - | 8 | 1281 | ||
AJ279792.1 | - | 7 | 1197 | ||
AJ279793.1 | - | 6 | 1102 | ||
AJ279794.1 | - | 6 | 1095 | ||
AJ279795.1 | - | 5 | 1000 | ||
AK013248.1 | - | 7 | 1012 | ||
AK154694.1 | - | 8 | 1342 | ||
AK171199.1 | - | 8 | 1426 | ||
AY039000.1 | - | 9 | 1214 | ||
BC051204.1 | - | 6 | 1125 | ||
BC055858.1 | - | 7 | 1211 | 4913671 |