Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_016474.4 | NP_057558.3 | 2 | 174 | |
NR_135523.1 | 2 | 174 | |||
XM_017006554.1 | XP_016862043.1 | 2 | 237 | ||
XM_017006555.1 | XP_016862044.1 | 2 | 235 | ||
GenBank mRNA | AF151046.1 | - | 2 | 139 | |
AK091158.1 | - | 2 | 168 | ||
AK309525.1 | - | 2 | 147 | ||
BC005199.1 | - | 2 | 158 | ||
BC013999.2 | - | 2 | 150 | 3936914 | |
BC033897.1 | - | 2 | 161 | 4812977 | |
BF698254.1 | - | 159 | |||
BP195134.1 | - | 2 | 174 | ||
CD580179.2 | - | 2 | 172 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
GenBank mRNA | BC012553.1 | - | 162 |