Gene Transcripts

Gene Symbol
KIZ
Entrez Gene ID
55857
Gene Name
kizuna centrosomal protein
Gene Aliases
C20orf19, HT013, Kizuna, NCRNA00153, PLK1S1, RP69
Chromosome Location
Chr. 20: 21125980 - 21246628 on Build GRCh38
UniGene ID
Hs.187635
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163022.1 NP_001156494.1 5 1237
NM_001163023.1 NP_001156495.1 4 1147
NM_001276389.1 NP_001263318.1 4 1174
NM_018474.4 NP_060944.3 6 1400
XM_011529296.2 XP_011527598.1 6 1403
XM_011529297.2 XP_011527599.1 6 1392
XM_011529298.2 XP_011527600.1 6 1385
XM_011529299.2 XP_011527601.1 5 1232
XM_017027951.1 XP_016883440.1 7 1346
XM_017027952.1 XP_016883441.1 5 1232
XM_017027953.1 XP_016883442.1 7 1418
XM_017027954.1 XP_016883443.1 5 1232
GenBank mRNA AF220187.1 - 2 432
AF451990.1 - 6 1377
AK025476.1 - 6 1331
AK293166.1 - 4 1078
AK293567.1 - 4 1118
AK295355.1 - 5 1136
AK296399.1 - 2 666
AL110196.1 - 2 679
BC039296.1 - 6 1361
BC045826.1 - 5 1124 5262165
BC065550.1 - 4 981
BC090879.2 - 2 805
BC105093.1 - 6 1303 8144096
BC113370.1 - 6 1303 8322422

View Full Product Details