Gene Transcripts

Gene Symbol
KIAA1456
Entrez Gene ID
57604
Gene Name
KIAA1456
Gene Aliases
C8orf79, TRM9L
Chromosome Location
Chr. 8: 12945674 - 13029777 on Build GRCh38
UniGene ID
Hs.202521
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099677.1 NP_001093147.1 2 1266
NM_020844.2 NP_065895.2 5 1556
XM_005273584.4 XP_005273641.1 5 1567
XM_005273585.4 XP_005273642.1 6 1578
XM_005273586.4 XP_005273643.1 6 1612
XM_005273587.4 XP_005273644.1 5 1800
XM_005273588.4 XP_005273645.1 6 1724
XM_005273590.4 XP_005273647.1 4 1256
XM_005273591.4 XP_005273648.1 5 1740
XM_011544596.2 XP_011542898.1 5 2027
XM_011544597.2 XP_011542899.1 5 1379
XM_011544598.2 XP_011542900.1 5 1648
XM_011544600.2 XP_011542902.1 5 1386
XM_011544601.2 XP_011542903.1 5 1509
XM_017013705.1 XP_016869194.1 5 1417
XM_017013706.1 XP_016869195.1 5 1638
XM_017013707.1 XP_016869196.1 6 1783
XM_017013708.1 XP_016869197.1 5 1882
XM_017013709.1 XP_016869198.1 6 1767
XM_017013710.1 XP_016869199.1 7 1726
XM_017013711.1 XP_016869200.1 6 1561
XM_017013712.1 XP_016869201.1 6 1938
XM_017013713.1 XP_016869202.1 5 1682
XM_017013714.1 XP_016869203.1 4 1484
XM_017013715.1 XP_016869204.1 5 2357
GenBank mRNA AB040889.1 - 2 1330
AK307825.1 - 2 1582
BC016633.2 - 2 909

View Full Product Details