Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001024372.1 | NP_001019543.1 | 2 | 942 | |
NM_024812.2 | NP_079088.1 | 3 | 1109 | ||
GenBank mRNA | AK022077.1 | - | 3 | 1087 | |
AK093819.1 | - | 2 | 795 | ||
AK310816.1 | - | 2 | 842 | ||
BC035038.2 | - | 2 | 922 |