Gene Transcripts
- Gene Symbol
- FAAP100
- Entrez Gene ID
- 80233
- Gene Name
- Fanconi anemia core complex associated protein 100
- Gene Aliases
- C17orf70
- Chromosome Location
- Chr. 17: 81539885 - 81552403 on Build GRCh38
- UniGene ID
- Hs.313905
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_025161.5 |
NP_079437.5
|
4, 5 |
1481 |
|
NR_033338.1 |
|
4, 5 |
1677 |
|
XM_006722111.2 |
XP_006722174.1
|
4, 5 |
1476 |
|
XM_006722112.3 |
XP_006722175.1
|
4, 5 |
1627 |
|
XM_017025162.1 |
XP_016880651.1
|
4, 5 |
1502 |
|
XM_017025163.1 |
XP_016880652.1
|
4, 5 |
1398 |
|
GenBank mRNA |
BC008883.2 |
- |
4, 5 |
1650 |
|
BC117139.1 |
- |
2, 3 |
1001 |
40125690 |
BC117141.1 |
- |
2, 3 |
1001 |
40125692 |
BC143861.1 |
- |
2, 3 |
1001 |
|
DQ989324.1 |
- |
4, 5 |
1463 |
|
View Full Product Details