Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001142459.1 | NP_001135931.2 | 2 | 791 | |
NM_001142460.1 | NP_001135932.2 | 2 | 791 | ||
NM_080871.3 | NP_543147.2 | 2 | 595 | ||
GenBank mRNA | AF417920.1 | - | 2 | 793 | |
AK055536.1 | - | 2 | 501 | ||
AK125565.1 | - | 2 | 595 | ||
AK315114.1 | - | 2 | 621 | ||
BC126351.1 | - | 2 | 555 | 8992067 |