Gene Transcripts

Gene Symbol
TMCO5A
Entrez Gene ID
145942
Gene Name
transmembrane and coiled-coil domains 5A
Gene Aliases
TMCO5
Chromosome Location
Chr. 15: 37934607 - 37997541 on Build GRCh38
UniGene ID
Hs.179646
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_152453.3 NP_689666.2 8 681
XM_005254168.3 XP_005254225.1 8 662
XM_011521262.2 XP_011519564.1 8 662
XM_011521263.1 XP_011519565.1 8 662
XM_011521264.1 XP_011519566.1 8 662
GenBank mRNA AK093126.1 - 8 662
AK310517.1 - 8 667
BC029221.2 - 7 594 5167874
BI561792.1 - 8 687
BI826726.1 - 8 645

View Full Product Details