Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001010863.2 | NP_001010863.1 | 3 | 243 | |
NM_001288740.1 | NP_001275669.1 | 3 | 243 | ||
NM_001288741.1 | NP_001275670.1 | 3 | 243 | ||
NM_001288742.1 | NP_001275671.1 | 3 | 243 | ||
NM_001288743.1 | NP_001275672.1 | 3 | 243 | ||
XM_011539378.2 | XP_011537680.1 | 3 | 423 | ||
XM_011539379.1 | XP_011537681.1 | 3 | 423 | ||
XM_011539380.1 | XP_011537682.1 | 3 | 423 | ||
XM_011539382.1 | XP_011537684.1 | 3 | 423 | ||
XM_011539383.2 | XP_011537685.1 | 3 | 423 | ||
XM_011539384.1 | XP_011537686.1 | 3 | 423 | ||
XM_011539385.1 | XP_011537687.1 | 3 | 423 | ||
XM_017015792.1 | XP_016871281.1 | 3 | 423 | ||
XM_017015793.1 | XP_016871282.1 | 3 | 423 | ||
XM_017015794.1 | XP_016871283.1 | 3 | 423 | ||
XM_017015795.1 | XP_016871284.1 | 3 | 423 | ||
XM_017015796.1 | XP_016871285.1 | 3 | 423 | ||
XM_017015797.1 | XP_016871286.1 | 3 | 423 | ||
GenBank mRNA | AK303891.1 | - | 3 | 241 | |
AL524299.3 | - | 3 | 218 | ||
BC031641.1 | - | 3 | 350 | ||
BC037922.2 | - | 3 | 234 | ||
BC042857.1 | - | 3 | 234 | ||
BC047724.1 | - | 3 | 199 | ||
BC104438.1 | - | 3 | 205 | ||
BC104439.1 | - | 3 | 205 | ||
BU145920.1 | - | 3 | 155 | ||
DA366138.1 | - | 3 | 196 | ||
DC389158.1 | - | 3 | 244 | ||
DQ884399.1 | - | 3 | 179 |