Gene Transcripts
- Gene Symbol
- ADAM32
- Entrez Gene ID
- 203102
- Gene Name
- ADAM metallopeptidase domain 32
- Gene Aliases
- Chromosome Location
- Chr. 8: 39107529 - 39284917 on Build GRCh38
- UniGene ID
- Hs.521545
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001313994.1 |
NP_001300923.1
|
3, 4 |
451 |
|
NM_145004.6 |
NP_659441.3
|
4, 5 |
531 |
|
XM_006716298.2 |
XP_006716361.1
|
4, 5 |
325 |
|
XM_006716299.1 |
XP_006716362.1
|
4, 5 |
325 |
|
XM_011544433.1 |
XP_011542735.1
|
4, 5 |
325 |
|
XM_011544434.1 |
XP_011542736.1
|
4, 5 |
325 |
|
XM_017013183.1 |
XP_016868672.1
|
4, 5 |
325 |
|
XM_017013184.1 |
XP_016868673.1
|
4, 5 |
325 |
|
XM_017013185.1 |
XP_016868674.1
|
3, 4 |
331 |
|
XM_017013186.1 |
XP_016868675.1
|
4, 5 |
325 |
|
GenBank mRNA |
AB209690.1 |
- |
4, 5 |
312 |
|
AK122608.1 |
- |
4, 5 |
325 |
|
AK300148.1 |
- |
3, 4 |
409 |
|
AK313864.1 |
- |
4, 5 |
352 |
|
AY172952.1 |
- |
4, 5 |
453 |
|
AY358739.1 |
- |
4, 5 |
411 |
|
BC026169.2 |
- |
4, 5 |
342 |
4829390 |
BC028702.2 |
- |
4, 5 |
325 |
4829385 |
BC030014.2 |
- |
4, 5 |
318 |
4830659 |
BC030698.2 |
- |
4, 5 |
333 |
4822988 |
BC034975.2 |
- |
4, 5 |
326 |
4829806 |
BI559590.1 |
- |
4, 5 |
536 |
|
View Full Product Details