Gene Transcripts

Gene Symbol
LMNTD2
Entrez Gene ID
256329
Gene Name
lamin tail domain containing 2
Gene Aliases
C11orf35
Chromosome Location
Chr. 11: 554850 - 560780 on Build GRCh38
UniGene ID
Hs.669395
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173573.2 NP_775844.2 6 655
XM_011519965.2 XP_011518267.2 6 830
XM_011519967.2 XP_011518269.2 6 830
XM_017017476.1 XP_016872965.1 6 830
XM_017017477.1 XP_016872966.1 6 815
XM_017017478.1 XP_016872967.1 6 671
XM_017017479.1 XP_016872968.1 6 830
GenBank mRNA BC039077.1 - 6 655
BX384085.2 - 7 758

View Full Product Details