Gene Transcripts

Gene Symbol
C19orf35
Entrez Gene ID
374872
Gene Name
chromosome 19 open reading frame 35
Gene Aliases
Chromosome Location
Chr. 19: 2274632 - 2282182 on Build GRCh38
UniGene ID
Hs.511803
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_198532.2 NP_940934.1 2 171
GenBank mRNA AK127680.1 - 2 165
BC107678.1 - 2 171

View Full Product Details