Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001005353.2 | NP_001005353.1 | 6 | 952 | |
NM_013410.3 | NP_037542.1 | 5 | 835 | ||
NM_203464.2 | NP_982289.1 | 6 | 841 | ||
XM_017000612.1 | XP_016856101.1 | 5 | 587 | ||
XM_017000613.1 | XP_016856102.1 | 5 | 773 | ||
GenBank mRNA | AK025926.1 | - | 5 | 799 | |
AK309057.1 | - | 5 | 543 | ||
AK313611.1 | - | 5 | 799 | ||
BC016180.1 | - | 5 | 735 | 4637988 | |
BC040224.1 | - | 5 | 711 | 5188877 | |
BC066944.1 | - | 6 | 952 | 4819382 | |
BC136886.1 | - | 5 | 676 | 9020877 | |
BC136887.1 | - | 1 | 676 | 9020878 | |
BC146653.1 | - | 6 | 826 | 40147793 | |
BC148270.1 | - | 5 | 616 | 8860213 | |
CR456830.1 | - | 5 | 594 | ||
X60673.1 | - | 5 | 777 |