Gene Transcripts

Gene Symbol
ALG2
Entrez Gene ID
85365
Gene Name
ALG2, alpha-1,3/1,6-mannosyltransferase
Gene Aliases
CDGIi, CMS14, CMSTA3, NET38, hALPG2
Chromosome Location
Chr. 9: 99216425 - 99221964 on Build GRCh38
UniGene ID
Hs.40919
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_033087.3 NP_149078.1 2 2306
NR_024532.1 3 2465
GenBank mRNA AK074988.1 - 3 2443
AK075189.1 - 1 1526

View Full Product Details