Gene Transcripts

Gene Symbol
ARX
Entrez Gene ID
170302
Gene Name
aristaless related homeobox
Gene Aliases
CT121, EIEE1, ISSX, MRX29, MRX32, MRX33, MRX36, MRX38, MRX43, MRX54, MRX76, MRX87, MRXS1, PRTS
Chromosome Location
Chr. X: 25003694 - 25015948 on Build GRCh38
UniGene ID
Hs.300304
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_139058.2 NP_620689.1 5 2390
GenBank mRNA CA775911.1 - 1 569

View Full Product Details