Gene Transcripts

Gene Symbol
CARD9
Entrez Gene ID
64170
Gene Name
caspase recruitment domain family member 9
Gene Aliases
CANDF2, hCARD9
Chromosome Location
Chr. 9: 136363956 - 136373681 on Build GRCh38
UniGene ID
Hs.694071
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_052813.4 NP_434700.2 13 2077
NM_052814.3 NP_434701.1 12 1768
GenBank mRNA AK024001.1 - 14 2131
AK292081.1 - 13 2029
BC008877.2 - 12 1743 4139529

View Full Product Details