Gene Transcripts

Gene Symbol
CCDC168
Entrez Gene ID
643677
Gene Name
coiled-coil domain containing 168
Gene Aliases
C13orf40
Chromosome Location
Chr. 13: 102729367 - 102759072 on Build GRCh38
UniGene ID
Hs.368382
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146197.1 NP_001139669.1 4 2358
XM_011521106.1 XP_011519408.1 5 2231

View Full Product Details